missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-b]pyridazine, 98%

Product Code. 15437908
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437908 25 g 25g
15427908 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437908 Supplier Thermo Scientific Alfa Aesar Supplier No. H61377.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 766-55-2 | C6H5N3 | 119.127 g/mol

Imidazo[1,2-b]pyridazine is used as an intermediate of cefozopran, building block and in fused ring systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazo[1,2-b]pyridazine is used as an intermediate of cefozopran, building block and in fused ring systems.

Solubility
Soluble in dimethylformamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-55-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD07782103
InChI Key VTVRXITWWZGKHV-UHFFFAOYSA-N
Synonym imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
PubChem CID 136599
IUPAC Name imidazo[1,2-b]pyridazine
SMILES C1=CC2=NC=CN2N=C1

Specifications

Melting Point 54°C
Quantity 25 g
Solubility Information Soluble in dimethylformamide.
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material Imidazo[1,2-b]pyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.