missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4-carboxaldehyde, 97%

Product Code. 11496017
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11496017 1 g 1g
11406027 5 g 5g
11416027 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11496017

Brand: Thermo Scientific Alfa Aesar B25050.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3034-50-2 | C4H4N2O | 96.089 g/mol

Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

Solubility
Soluble in DMSO, methanol.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-50-2
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00173726
InChI Key ZQEXIXXJFSQPNA-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carbaldehyde, imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, imidazole-4-carboxaldehyde, 4-imidazolecarboxaldehyde, 4-formylimidazole, 4 5-imidazolecarboxaldehyde, 3h-imidazole-4-carbaldehyde, 4-formyl-1h-imidazole, imidazole-5-carbaldehyde
PubChem CID 76428
IUPAC Name 1H-imidazole-5-carbaldehyde
SMILES C1=C(NC=N1)C=O

Specifications

Melting Point 174°C to 177°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO,methanol.
Formula Weight 96.09
Percent Purity 97%
Chemical Name or Material Imidazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.