missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, 99+%, crystalline

Product Code. 10697821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697821 250 g Plastic bottle 250g
10561331 1 kg Plastic bottle 1kg
10522714 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10697821 Supplier Thermo Scientific Acros Supplier No. 301872500
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Imidazole, 99+%, C3H4N2, CAS Number-288-32-4, glyoxalin, imidazole, miazole, imutex, pyrro b monazole, 1,3-diazole, imidazol, iminazole, 1,3-diaza-2,4-cyclopentadiene, glyoxaline, 1kg, 99% min. (GC), 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow or Beige, 206-019-2, 01,492; 02,220 | CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 86.0°C to 90.0°C
Color White to Yellow or Beige
Boiling Point 255.0°C to 256.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 23, 45
Fieser 01,492; 02,220
Merck Index 15, 4953
Solubility Information Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether
Formula Weight 68.08
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Imidazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.