missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole, Extra Pure, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Specifications
Specifications
| Melting Point | 86°C |
| Color | White to Yellow |
| pH | 10.5 |
| Boiling Point | 255°C |
| Packaging | Plastic powder jar |
| Quantity | 1 kg |
| Formula Weight | 68.08g/mol |
| Vapor Pressure | 0.003mbar at 20°C |
| Physical Form | Solid |
Safety and Handling
Product Identifier
- Imidazole
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction