missing translation for 'onlineSavingsMsg'
Learn More

Imidazole (Molecular Biology), Fisher BioReagents™,50g

Product Code. 10522765
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly Bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10522765 50 g Poly Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10522765 Supplier Fisher BioReagents Supplier No. BP30550
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Commonly used buffering agent

Imidazole is an excellent biological buffer with a usable pH range of 6.2 to 7.8.
TRUSTED_SUSTAINABILITY

Specifications

CAS 288-32-4
Absorbance 0.01 max. (0.1M solution) at 260nm
Melting Point 86°C
Color White to Yellow
pH 10.5
Boiling Point 255°C
Assay Percent Range ≥99 %
Packaging Poly Bottle
Molecular Formula C3H4N2
Linear Formula HNCH:CHN:CH
MDL Number MFCD00005183
Quantity 50 g
Merck Index 15, 4953
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.08
PubChem CID 795
ChEBI CHEBI:16069
Formula Weight 68.08
Vapor Pressure 0.003mbar at 20°C
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material Imidazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.