missing translation for 'onlineSavingsMsg'
Learn More

Imidazolidinyl urea

Product Code. 15467218
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467218 25 g 25g
15477218 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467218 Supplier Thermo Scientific Alfa Aesar Supplier No. H60729.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39236-46-9 | C11H16N8O8 | 388.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39236-46-9
Molecular Formula C11H16N8O8
Molecular Weight (g/mol) 388.297
MDL Number MFCD00038890
InChI Key ZCTXEAQXZGPWFG-UHFFFAOYSA-N
Synonym imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
PubChem CID 38258
ChEBI CHEBI:51805
IUPAC Name 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
SMILES C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO

Specifications

Melting Point 141°C to 143°C
Quantity 25 g
Formula Weight 388.3
Chemical Name or Material Imidazolidinyl urea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.