missing translation for 'onlineSavingsMsg'
Learn More

Imidurea, TRC

Product Code. 30408378
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408378 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408378 Supplier Toronto Research Chemicals Supplier No. TRCI3875705G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imidurea
CAS 39236-46-9
Molecular Formula C11 H16 N8 O8
Quantity 5 g
Synonym Abiol, Biopure 100, Euxyl K 200, Germall 115, Imidazolidinyl urea, Imidurea, Kemipur 100, N,N''-Methylenebis[N'-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]urea], Sepicide CI, Urea, N,N''-methylenebis[N'-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-
InChI Formula InChI=1S/C11H16N8O8/c20-2-18-4(6(22)16-10(18)26)14-8(24)12-1-13-9(25)15-5-7(23)17-11(27)19(5)3-21/h4-5,20-21H,1-3H2,(H2,12,14,24)(H2,13,15,25)(H,16,22,26)(H,17,23,27)
SMILES OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O
IUPAC Name 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
Molecular Weight (g/mol) 388.29
Formula Weight 388.1091
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.