missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iminodiacetic acid, 98+%

Product Code. 10711291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Unit Size:
2.5kg
25g
500g
Product Code. Quantity unitSize
10711291 500 g 500g
10256640 25 g 25g
10626594 2.5 kg 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 10711291

Brand: Thermo Scientific Chemicals 204975000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-73-4 | C4H7NO4 | 133.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.1
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O

Specifications

Melting Point 243°C
Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HN(CH2CO2H)2
Quantity 500 g
Beilstein 04, 365
Merck Index 15, 4959
Solubility Information Solubility in water: 2.43g/100 mL water (5°C). Other solubilities: 2.9g/100 mL water (20°C),6.3g/100 mL water (40°C),12.8g/100 mL water (60°C),23.0g/100 mL water (80°C),32.5g/100 mL water (100°C)
Formula Weight 133.1
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Iminodiacetic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Iminodiacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Iminodiacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.