missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetic Acid, TRC

Product Code. 30138773
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138773 5 g 5g
30010762 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138773 Supplier Toronto Research Chemicals Supplier No. TRCI4080505G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4 H7 N O4
Molecular Weight (g/mol) 133.1
Synonym N-(carboxymethyl)-Glycine, Acetic acid, iminodi- (6CI,8CI), ((Carboxymethyl)amino)acetic acid, Acetic acid, 2,2'-iminobis-, Aminodiacetic acid, Diglycin, Diglycine, Diglycocoll, IDA, IDA (chelating agent), IMDA, Iminobis(acetic acid), Iminodiacetic acid, Iminodiethanoic acid, N-(Carboxymethyl)glycine, NSC 18467, α-Iminodiacetic acid
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES OC(=O)CNCC(=O)O

Specifications

InChI Formula InChI=1S/C4H7NO4/c6-3(7)1-5-2-4(8)9/h5H,1-2H2,(H,6,7)(H,8,9)
Quantity 5 g
Formula Weight 133.0375
Shipping Condition Room Temperature
Chemical Name or Material Iminodiacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.