missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetonitrile, 95%

Product Code. 11420068
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420068 2500 g 2500g
11400068 100 g 100g
11410068 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420068 Supplier Thermo Scientific Alfa Aesar Supplier No. B23660.0E
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-87-5 | C4H5N3 | 95.11 g/mol

Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

Solubility
Soluble in water, acetone and hot methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-87-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00001887
InChI Key BSRDNMMLQYNQQD-UHFFFAOYSA-N
Synonym iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
PubChem CID 69413
ChEBI CHEBI:71189
IUPAC Name 2-(cyanomethylamino)acetonitrile
SMILES N#CCNCC#N

Specifications

Melting Point 75°C to 79°C
Quantity 2500 g
Beilstein 773803
Solubility Information Soluble in water,acetone and hot methanol.
Formula Weight 95.11
Percent Purity 95%
Chemical Name or Material Iminodiacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.