missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene, 97%

Product Code. 10041920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041920 10 g Glass bottle 10g
10563141 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041920 Supplier Thermo Scientific Acros Supplier No. 162540100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 256-96-2 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256-96-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00005071
InChI Key LCGTWRLJTMHIQZ-UHFFFAOYSA-N
Synonym iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino
PubChem CID 9212
ChEBI CHEBI:47802
IUPAC Name 11H-benzo[b][1]benzazepine
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2

Specifications

Melting Point 197°C to 201°C
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 193.25
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Iminostilbene, 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.