missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene, 97%

Product Code. 11479237
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479237 500 g 500g
11459237 25 g 25g
11469237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479237 Supplier Thermo Scientific Alfa Aesar Supplier No. B21323.36
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 256-96-2 | C14H11N | 193.249 g/mol

Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.

Solubility
Soluble in ethyl acetate (25 mg/ml), water (partly), dioxane, chloroform, and DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256-96-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00005071
InChI Key LCGTWRLJTMHIQZ-UHFFFAOYSA-N
Synonym iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino
PubChem CID 9212
ChEBI CHEBI:47802
IUPAC Name 11H-benzo[b][1]benzazepine
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2

Specifications

Melting Point 197°C to 199°C
Quantity 500 g
Beilstein 1343358
Solubility Information Soluble in ethyl acetate (25mg/ml),water (partly),dioxane,chloroform,and DMSO.
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material Iminostilbene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.