missing translation for 'onlineSavingsMsg'
Learn More

Imipenem, Monohydrate, TRC

Product Code. 30010583
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010583 50 mg 50mg
17991505 10 mg 10mg
17990925 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010583 Supplier Toronto Research Chemicals Supplier No. TRCI46520050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74431-23-5
Molecular Formula C12 H17 N3 O4 S . H2 O
Molecular Weight (g/mol) 317.36
Synonym Imipenem monohydrate, (5R,6S)-6-[(R)-1-Hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]sulfanyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid monohydrate, 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, (5R,6S)-6-[(1R)-1-hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-, hydrate (1:1), 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 6-(1-hydroxyethyl)-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-, monohydrate, [5R-[5α,6α(R*)]]-, 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 6-[(1R)-1-hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-, monohydrate, (5R,6S)- (9CI), Imipenem monohydrate, N-Formimidoyl thienamycin monohydrate
IUPAC Name (5R,6S)-6-[(1R)-1-hydroxyethyl]-3-(2-methanimidamidoethylsulfanyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
SMILES O.C[C@@H](O)[C@@H]1[C@H]2CC(=C(N2C1=O)C(=O)O)SCCNC=N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H17N3O4S.H2O/c1-6(16)9-7-4-8(20-3-2-14-5-13)10(12(18)19)15(7)11(9)17;/h5-7,9,16H,2-4H2,1H3,(H2,13,14)(H,18,19);1H2/t6-,7-,9-;/m1./s1
Formula Weight 317.1045
Percent Purity >95%
Chemical Name or Material Imipenem monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.