missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Imipramine hydrochloride
Description
Imipramine hydrochloride is a potent tricyclic antidepressant; inhibits 5-HT and noradrenalin transporters (Ki values = 1.3-20 nM and 20-142 nM respectively). Increases autophagy, both alone and in combination with VEGF inhibitors or Temozolomide (Cat. No. 2706), to reduce the progression of glioma and slow the proliferation of other tumors. Reprograms tumor-associated macrophages from immunosuppressive to immunostimulatory via histamine receptor inhibition.
Specifications
Specifications
| Chemical Name or Material | 5-(3-Dimethylaminopropyl)-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride |
| CAS | 113-52-0 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Autophagy-related Compounds |
| Molecular Formula | C19H24N2.HCl |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?