Learn More
Imipramine hydrochloride, Thermo Scientific™
A serotonin uptake inhibitor
65.00 € - 207.00 €
Chemical Identifiers
| CAS | 113-52-0 |
|---|---|
| Molecular Formula | C19H25ClN2 |
| Molecular Weight (g/mol) | 316.873 |
| MDL Number | MFCD00012669 |
| InChI Key | XZZXIYZZBJDEEP-UHFFFAOYSA-N |
| Synonym | imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram |
| PubChem CID | 8228 |
| ChEBI | CHEBI:5882 |
| IUPAC Name | 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride |
| SMILES | CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15475909
|
Thermo Scientific Alfa Aesar
J63723.06 |
5g |
65.00 €
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15485909
|
Thermo Scientific Alfa Aesar
J63723.14 |
25g |
207.00 €
25g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsImipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.
Solubility
Soluble in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 113-52-0 | |
| 316.873 | |
| XZZXIYZZBJDEEP-UHFFFAOYSA-N | |
| 8228 | |
| 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride |
| C19H25ClN2 | |
| MFCD00012669 | |
| imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram | |
| CHEBI:5882 | |
| CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl |
Specifications
| 113-52-0 | |
| MFCD00012669 | |
| imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram | |
| XZZXIYZZBJDEEP-UHFFFAOYSA-N | |
| 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride | |
| 8228 | |
| 316.87 |
| C19H25ClN2 | |
| 14,4920 | |
| Soluble in water | |
| CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl | |
| 316.873 | |
| CHEBI:5882 | |
| Imipramine hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 204-030-7
missing translation for 'rtecsNumber' : HO1925000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.