missing translation for 'onlineSavingsMsg'
Learn More

Imipramine hydrochloride, Thermo Scientific™

A serotonin uptake inhibitor

Brand:  Thermo Scientific Alfa Aesar J63723.06

Quantity 5g

 View more versions of this product

Product Code. 15475909

  • 65.00 € / 5g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

113-52-0
316.873
XZZXIYZZBJDEEP-UHFFFAOYSA-N
8228
3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
C19H25ClN2
MFCD00012669
imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
CHEBI:5882
CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

113-52-0
MFCD00012669
14,4920
Soluble in water
CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl
316.873
CHEBI:5882
Imipramine hydrochloride
C19H25ClN2
5g
imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
XZZXIYZZBJDEEP-UHFFFAOYSA-N
3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
8228
316.87
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.