missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Alum Adjuvant

Product Code. 10475325
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10475325 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10475325 Supplier Thermo Scientific™ Supplier No. 77161

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Stimulate a nonspecific immune response for mixed antigens (prepared immunogens) using this formulation of aluminum hydroxide and magnesium hydroxide.

Thermo Scientific™ Imject Alum Adjuvant is a formulation of aluminum hydroxide and magnesium hydroxide that effectively stimulates the immune response for antibody production procedures when mixed and injected with prepared immunogens.

Alum is considerably easier to mix with immunogens than Freund's adjuvants, as it does not require laborious emulsification. Although its effects are less pronounced compared to Freund's complete adjuvant (FCA), Imject Alum produces sufficient stimulation when used with complex immunogens (i.e., those that have been prepared as conjugates with KLH or other immunogenic carrier proteins).

Highlights:

  • Suspension of aluminum hydroxide and magnesium hydroxide
  • Mild but effective adjuvant for substantial immunogens
  • Ready to use: mix antigen directly with Imject Alum and inject
  • The adjuvant of choice for allergy and immunogenicity testing with or without sensitization and challenge with fraction V ovalbumin
  • Disadvantage: does not elicit as strong an immune response as FCA; requires a strong immunogen
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at room temperature. Product shipped at ambient temperature.

Note: This product is supplied for research or manufacturing applications only and is not intended for clinical use.
Quantity 50 mL
Product Line Imject
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.