missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Immethridine dihydrobromide, Tocris Bioscience™
Description
Potent and highly selective histamine H3 receptor agonist (pEC50 = 9.74) that displays 300-fold selectivity over the H4 receptor (pKi values are 9.07 and 6.61 respectively). Does not bind to H1 or H2 receptors at concentrations up to 10 μM. .
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 321.01 |
| Percent Purity | >99% |
| Chemical Name or Material | Immethridine dihydrobromide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?