Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Product Code. 10391315 Shop All Cytiva Products
Click to view available options
Type:
Anode
Cathode
Dry Strip Slinger
Immobiline DryStrip Kit
Sample Cup
Sample Cup Bar
Tray and Electrode Holder
Unit Size:
1 set
Each
Pack of 4
Pack of 60
Product Code. 10391315
Brand: Cytiva 18101867
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
  • The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
  • Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
  • Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
  • Individual components are also available separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Cathode