missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™-E Transfer Membrane

Product Code. 15867541 Shop All Merck Products
Change view
Click to view available options
Quantity:
1 Roll
10 Sheets
4 Sheets
50 Sheets
Dimensions (LxW):
1.875 m x 26.5 cm
10 m x 8.5 cm
10 x 10 cm
7 x 8.4 cm
8 x 10 cm
9 x 12 cm
Unit Size:
1 roll
Each
Pack of 10
Pack of 4
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dimensions (LxW) unitSize
15867541 1 Roll 1.875 m x 26.5 cm 1 roll
16379591 1 Roll 10 m x 8.5 cm Each
15897531 4 Sheets 7 x 8.4 cm Pack of 4
15807541 50 Sheets 7 x 8.4 cm Pack of 50
15817541 10 Sheets 8 x 10 cm Pack of 10
15827541 10 Sheets 9 x 12 cm Pack of 10
15837541 10 Sheets 10 x 10 cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15867541 Supplier Merck Supplier No. IEVH00005
  • ONLINE EXCLUSIVE PRICE
    313.00 €
    289.68 € / 1 rolls
    Save 23.32 €
    7% Off
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step.

Features:

  • 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
  • Saves time and reduces waste by eliminating the alcohol pre-wet step
  • Higher protein retention delivers stronger signal than most nitrocellulose membranes
TRUSTED_SUSTAINABILITY

Specifications

Description Easily wetted with water
For Use With (Application) Western Blotting
Color White
Code IEVH
Surface Plain
Quantity 1 Roll
Format Roll
Material PVDF
Pore Size 0.45 μm
Dimensions (LxW) 1.875 m x 26.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.