missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon-NC Membranes, Triton-Free, 0.45 μm

Product Code. 10166480 Shop All Merck Products
Change view
Click to view available options
Diameter (Metric):
82 mm
85 mm
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10166480 85 mm Pack of 50
10238200 82 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10166480 Supplier Merck Supplier No. HATF08550
  • ONLINE EXCLUSIVE PRICE
    258.00 €
    238.63 € / Pack of 50
    Save 19.37 €
    7% Off
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mixed Cellulose Esters, Surfactant-free

0.45 μm (0.45 micron filter) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color White
Material MCE
Pore Size 0.45 μm
Sterility Non-sterile
Flow Rate 60 mL/min x cm2
For Use With (Application) Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.).
Membrane Type Plain
Type Blotting Membranes
Wettability Hydrophilic
Diameter (Metric) 85 mm
Show More Show Less
compliance-icons
Product Identifier
  • Immobilon-NC Triton-free 85mm disc
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.