missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™-P Transfer Membrane

Product Code. 11700433 Shop All Merck Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11700433 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11700433 Supplier Merck Supplier No. IPVH22220

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Immobilon PVDF membranes provide many advantages compared to nitrocellulose. They won't crack or curl, and can be cut without fracturing. They also have low background, broad solvent compatibility and superior standing capabilities. Excellent protein retention and high physical strength.

  • Recommended for Western blotting
  • Convenient blotting sandwiches feature pre-cut sheets of membrane
  • High protein adsorption - no loss of proteins during transfer or reprobing
  • Open pore structure makes it easy to access bound proteins and remove unbound probes
  • Optimised for fluorescent blots to dramatically increase signal-to-noise ratios for high sensitivity in quantitative, multiplexing applications
  • Won't crack or curl
  • Low background
  • Superior staining capabilities
  • Can be reprobed multiple times
  • Applications: High Sensitivity Immunodetection, Binding Assays, Dot/slot Blotting, Glycoprotein Visualization, Lipopolysaccharide Analysis, Mass Spectrometry, Amino Acid Analysis, N-terminal Protein Sequencing
TRUSTED_SUSTAINABILITY

Spezifikation

For Use With (Application) High Sensitivity Immunodetection, Binding Assays, Dot/slot Blotting, Glycoprotein Visualization, Lipopolysaccharide Analysis, Mass Spectrometry, Amino Acid Analysis, N-terminal Protein Sequencing
Binding Capacity Insulin: 160 μg/cm2, Bovine Serum Albumin (BSA): 215 μg/cm2, Goat IgG: 294 μg/cm2
Color White
Format Sheet
Material PVDF
Pore Size 0.45 μm
Dimensions (LxW) 222 mm x 222 mm
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.