missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Immu-Mount™

Product Code. 10662815 Shop All Epredia Products
Change view
Click to view available options
Quantity:
20 mL
6 x 20 mL
1 L
Unit Size:
20mL
Pack of 2
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10662815 20 mL 20mL
12569996 1 L Pack of 2
10622689 6 x 20 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10662815 Supplier Epredia™ Supplier No. 9990402
  • ONLINE EXCLUSIVE PRICE
    30.55 €
    23.71 € / 20mL
    Save 6.84 €
    22% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-permanent mountant for use with frozen sectioning, immunoperoxidase techniques and immunofluorescent applications.

  • Economical, organic, overnight-drying liquid coverslip
  • Aqueous base for immunostaining and other techniques
  • Non-fluorescing medium with a low viscosity
  • Resists fading and polymerizes quickly for easier handling of finished slides
  • Refractive index: 1.495 ±0.002 (liquid); 1.586 ± 0.002 (dry)
  • Viscosity: Low (120-160 cps at 24°C)
  • pH: 8.0 from 8.8 at 21 through 24°C
TRUSTED_SUSTAINABILITY

Specifications

Applications Frozen section, immunoperoxidase techniques, and immunofluorescent preparations
Quantity 20 mL
Product Type Shandon Immu-mount
Volume (Metric) 20 mL
compliance-icons
Product Identifier
  • shandon mount
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: toluene, butyl methacylate, 2,6-di-tert-butyl-p-cresol, butyl benzyl phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.