missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Plex Human Immune Response Panel, 80-plex

Product Code. 32045521
Change view
Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
Unit Size:
Each
Pack of 10
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32045521 1 Plate Each
32044889 3 Plates Pack of 3
32044426 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32045521 Supplier Invitrogen™ Supplier No. QGP18010080
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The QuantiGene Plex Human Immune Response Panel, 80-plex, enables the study of immune function by analyzing 80 RNA targets and biomarkers in a single well using Luminex xMAP technology in combination with branched DNA signal amplification.

The QuantiGene Plex Human Immune Response Panel, 80-plex, enables the study of immune function by analyzing 80 RNA targets and biomarkers in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. Analyze 80 cytokine, chemokine, and growth factor targets simultaneously for efficient immune response profiling, biomarker discovery, and validation.

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide a high level of validation and precision. This kit is not compatible with the MAGPIX instrument.

Features of the QuantiGene Plex Human Immune Response Panel, 80-plex include:

  • True multiplexing—measure 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-plate format compatible with Luminex 200, FLEXMAP3D, and INTELLIFLEX systems; 384-well format available on demand
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targets—if alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to create pathway and disease-themed custom panels
  • Rapid turnaround for this catalogue panel for fast delivery

Targets (80-plex): CCL1, CCL13, CCL17 ,CCL19, CCL2, CCL21, CCL22, CCL23, CCL24, CCL25, CCL26, CCL4, CCL7, CCL8, CD40LG, CSF1, CSF2, CSF3, CX3CL1, CXCL1, CXCL11, CXCL13, CXCL2, CXCL5, CXCL6, CXCL9, CXCR3, FGF2, GZMA, GZMB, HGF, IFNA1, IFNG, IL10, IL12A, IL12B, IL13, IL15, IL16, IL17A, IL18, IL1A, IL1B, IL2, IL20, IL21, IL22, IL23A, IL27, IL2RA, IL3, IL31, IL34, IL37, IL4, IL5, IL6, IL8, IL9, KITLG, LGALS3, LIF, LTA, MIF, NGF, PTX3, TNF, TNFRSF12A, TNFRSF1B, TNFRSF8, TNFSF10, TNFSF13, TNFSF13B, TREM1, TSLP, VEGFA, PPIB, HPRT1, GAPDH, GUSB

Panel and signal amplification technology

QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA directly from cell lysates, tissue homogenates, or purified RNA, and they enable the measurement of multiple targets in the same well via multiplexing. QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. They use the target-specific probes to capture the RNA of interest, and branched DNA technology then amplifies fluorescent signals which are read with a Luminex xMAP instrument. The fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and with this specific panel enables human immune profiling. Detailed and step-by-step data analysis is facilitated via the QuantiGene Plex data analysis app.

Applications

QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology, and detection of translocations and fusion genes. In general, any multiplex gene expression analysis can be done with QuantiGene Plex panels, independent of species or sample type.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well
For Use With (Application) QuantiGene™ Gene Expression Assay
For Use With (Equipment) Luminex instrument
Content And Storage

Target-specific probe set and magnetic capture beads (premixed and ready to use).

Store probe set and capture beads at 4°C.

Do not freeze the capture beads, as they can be damaged if frozen.

Product Line QuantiGene™ Plex
Product Type Multiplex Panel
Quantity 1 Plate
Shipping Condition Wet Ice
Sample Type Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Detection Method Fluorescence
Research Area Immunology
Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.