missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ImmunoCult™-ACF Dendritic Cell Medium

Product Code. 17137651
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17137651 100 mL 100mL
16655635 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17137651 Supplier STEMCELL Technologies Supplier No. 10987
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation

  • ImmunoCult™-ACF Dendritic Cell Medium and Supplements, sold individually or as a kit, have been developed for the in vitro culture and differentiation of human monocytes into dendritic cells (DCs)
  • ImmunoCult™-ACF Dendritic Cell Medium is a serum-free and animal component-free medium optimized for the in vitro culture and differentiation of human monocytes into DCs
  • ImmunoCult™-ACF Dendritic Cell Differentiation Supplement contains a combination of animal component-free recombinant human cytokines formulated to support the differentiation of immature DCs from human monocytes
  • The ImmunoCult™ Dendritic Cell Maturation Supplement has been formulated to support maturation of immature DCs to mature DCs

Specifications

Cell Type Dendritic Cells, Monocytes
For Use With (Application) Cell Culture; Differentiation
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.