missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ImmunoCult™ Human CD3/CD28 T Cell Activator

Product Code. 16425234
Encompass
Change view
Click to view available options
Quantity:
2 mL
5 x 2 mL
Unit Size:
2mL
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16425234 5 x 2 mL Pack of 5
15553062 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16425234 Supplier STEMCELL Technologies Supplier No. 10991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human T cell activation and expansion reagent

  • Achieve robust activation and expansion of T cells in the absence of magnetic beads, feeder cells, or antigens
  • This product's gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded, for example in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981)
  • Tetrameric antibody complexes bind to and cross-link CD3 and CD28 cell surface ligands, thereby providing the required signals for T cell activation
  • ImmunoCult™ Human CD3/CD28 T Cell Activator can be used on the Seahorse XF Analyzer to measure T cell activation response and is also available as part of the Agilent Seahorse XF Hu T Cell Activation Assay Kit
  • This product is designed for cell therapy research applications, but may be qualified for use as an ancillary material (AM) following the framework outlined in USP<1043>
  • STEMCELL Technologies can work with you to qualify this reagent as an AM under an approved investigational new drug (IND) or clinical trial application (CTA)

Spezifikation

Cell Type T Cells, T Cells, CD4+, T Cells, CD8+
For Use With (Application) Activation, Cell Culture, Expansion
Quantity 5 x 2 mL
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.