Learn More
Tocris Bioscience™ IN 1130
GREENER_CHOICE
Potent and selective inhibitor of TGF-βRI
Brand:  Tocris Bioscience™ 6016/5
Encompass
Product Code. 15776349
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
IN 1130 is a potent and selective inhibitor of TGF-β type I receptor (ALK5). Inhibits Smad-2 phosphorylation (IC50 = 5.3 nM). Exhibits selectivity over a panel of 27 serine/ threonine and tyrosine kinases including P38α. Attenuates TGFβ/Smad signaling, cell migration, invasion, lung metastasis and increases survival of mice in 4TI in vivo models. Suppresses renal fibrosis in obstructive nephropathy in vivo models.
Specifications
Specifications
3-[[5-(6-Methyl-2-pyridinyl)-4-(6-quinoxalinyl)-1H-imidazol-2-yl]methyl]benzamide
868612-83-3
>98%