missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IN 1130
GREENER_CHOICE

Product Code. 18720318
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18720318 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18720318 Supplier Tocris Bioscience™ Supplier No. 6016/5
  • ONLINE EXCLUSIVE PRICE
    200.00 €
    189.00 € / 5mg
    Save 11.00 €
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective inhibitor of TGF-βRI

IN 1130 is a potent and selective inhibitor of TGF-β type I receptor (ALK5). Inhibits Smad-2 phosphorylation (IC50 = 5.3 nM). Exhibits selectivity over a panel of 27 serine/ threonine and tyrosine kinases including P38α. Attenuates TGFβ/Smad signaling, cell migration, invasion, lung metastasis and increases survival of mice in 4TI in vivo models. Suppresses renal fibrosis in obstructive nephropathy in vivo models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[[5-(6-Methyl-2-pyridinyl)-4-(6-quinoxalinyl)-1H-imidazol-2-yl]methyl]benzamide
CAS 868612-83-3
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C25H20N6O
Formula Weight Observed MW: 420.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.