missing translation for 'onlineSavingsMsg'
Learn More

Inconel∣r 600 gauze, 10 mesh woven from 0.64mm (0.025in) dia wire

Product Code. 15477215
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477215 300 x 300 mm Each
15457215 75 x 75 mm Each
15467215 150 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15477215 Supplier Thermo Scientific Alfa Aesar Supplier No. 046453.HB
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inconel is typically used in applications where high temperatures are present. Inconel superalloys also have excellent mechanical properties and are frequently used in the chemical and aerospace industries. It is common in gas turbine blades, seals, and as well as turbocharger rotors and seals, electric submersible well pump motor shafts, high temperature fasteners, chemical processing and pressure vessels, heat exchanger tubing, steam generators and core components in nuclear pressurized water reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Inconel™ 600 gauze
Name Note 10 mesh woven from 0.64mm (0.025 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr:Fe; 77:15.5:7.5 wt%
Quantity 300 x 300 mm
Mesh Size 10 Mesh
Form Wire Cloth
MDL Number MFCD01741927
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.