missing translation for 'onlineSavingsMsg'
Learn More

Indazole-3-carboxylic Acid, TRC

Product Code. 30393739
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393739 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393739 Supplier Toronto Research Chemicals Supplier No. TRCI50500010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Indazole-3-carboxylic Acid
CAS 4498-67-3
Molecular Formula C8 H6 N2 O2
Quantity 10 g
Synonym 1H-Indazole-3-carboxylic Acid, Granisetron Hydrochloride Imp. H (EP), Indazole-3-carboxylic acid (7CI), 3(1H)-Indazolecarboxylic acid, 3-Carboxy-1H-indazole, 3-Carboxyindazole, NSC 520610
InChI Formula InChI=1S/C8H6N2O2/c11-8(12)7-5-3-1-2-4-6(5)9-10-7/h1-4H,(H,9,10)(H,11,12)
SMILES OC(=O)c1n[nH]c2ccccc12
IUPAC Name 1H-indazole-3-carboxylic acid
Molecular Weight (g/mol) 162.145
Formula Weight 162.043
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.