missing translation for 'onlineSavingsMsg'
Learn More

Indazole, 96%

Product Code. 10112590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112590 5 g Glass bottle 5g
11916601 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10112590 Supplier Thermo Scientific Acros Supplier No. 122120050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 271-44-3 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-44-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
InChI Key BAXOFTOLAUCFNW-UHFFFAOYSA-N
Synonym indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
PubChem CID 9221
ChEBI CHEBI:36669
IUPAC Name 1H-indazole
SMILES C1=CC=C2C(=C1)C=NN2

Specifications

Melting Point 146.0°C to 150.0°C
Color Beige to White
Boiling Point 270.0°C (743.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 5 g
Beilstein 23,122
Merck Index 15,4976
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
Formula Weight 118.14
Percent Purity 96%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material Indazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.