missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo Carmine, for analysis, high purity biological stain

Product Code. 10734471 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10734471 500 g Glass bottle 500g
10724471 25 g Glass bottle 25g
10215680 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10734471 Supplier Thermo Scientific Chemicals Supplier No. 157445000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 860-22-0 | C16H8N2Na2O8S2 | 466.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt
PubChem CID 5284351
IUPAC Name disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Spezifikation

Dye Content 85% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Beilstein 25, 304
Merck Index 15, 4982
Color Blue
Solubility Information Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents
Quantity 500 g
Physical Form Fine Crystalline Powder
Formula Weight 466.34
Grade Analysis
Chemical Name or Material Indigo Carmine
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.