missing translation for 'onlineSavingsMsg'
Learn More

Indium, 99.99%, (trace metal basis), foil, 0.127 mm thick

Product Code. 11375436 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.3 g
9.2 g
Unit Size:
2.3g
9.2g
This item is not returnable. View return policy

Product Code. 11375436

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Gray
Melting Point 156.0°C
Boiling Point 2000.0°C
Quantity 9.2 g
Assay Percent Range 99.99%
Form Foil
Linear Formula In
Fieser 06,293; 14,181; 16,181; 17,152
Merck Index 15, 4987
Formula Weight 114.82
Percent Purity 99.99%
Grade Trace Metal Basis
Chemical Name or Material Indium
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.