missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) 2,4-pentanedionate, 98%

Product Code. 11351329
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11351329

Brand: Thermo Scientific Alfa Aesar 013794.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in catalysts | CAS: 14405-45-9 | C15H21InO6 | 412.15 g/mol

Used in catalysts.Indium(III) 2,4-pentanedionate is used as a precursor for the preparation of indium(III)oxide. It acts as a catalyst in the synthesis of oxindole derivatives. It finds application in gas sensors. Further, it is used in the synthesis of indium tin oxide nanocrystals. In addition to this, it is involved in the preparation of thin-film solar cells by atomic layer chemical vapor deposition with hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14405-45-9
Molecular Formula C15H21InO6
Molecular Weight (g/mol) 412.15
MDL Number MFCD00013494
InChI Key CMGVNFXMZKDHDG-UHFFFAOYSA-N
Synonym acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate
PubChem CID 101644361
IUPAC Name indium(3+);(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate
SMILES [In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 186°C (decomposition)
Quantity 5 g
Solubility Information Soluble in benzene; Insoluble in water.
Formula Weight 412.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Indium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.