missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) 2,4-pentanedionate, 98%

Product Code. 11341329
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341329 1 g 1g
11351329 5 g 5g
11361329 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11341329 Supplier Thermo Scientific Alfa Aesar Supplier No. 013794.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Used in catalysts | CAS: 14405-45-9 | C15H21InO6 | 412.15 g/mol

Used in catalysts.Indium(III) 2,4-pentanedionate is used as a precursor for the preparation of indium(III)oxide. It acts as a catalyst in the synthesis of oxindole derivatives. It finds application in gas sensors. Further, it is used in the synthesis of indium tin oxide nanocrystals. In addition to this, it is involved in the preparation of thin-film solar cells by atomic layer chemical vapor deposition with hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14405-45-9
Molecular Formula C15H21InO6
Molecular Weight (g/mol) 412.15
MDL Number MFCD00013494
InChI Key CMGVNFXMZKDHDG-UHFFFAOYSA-N
Synonym acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate
PubChem CID 101644361
IUPAC Name indium(3+);(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate
SMILES [In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 186°C (decomposition)
Quantity 1 g
Solubility Information Soluble in benzene; Insoluble in water.
Formula Weight 412.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Indium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.