Learn More
Indium(III) fluoride, anhydrous, 99.95% (metals basis)
Description
Indium(III) fluoride is used in the preparation of non-oxide glasses. It acts as a catalyst in the reaction of trimethylsilyl cyanide (TMSCN) to aldehydes to get cyanohydrins. It play a vital role as a catalyst in organic synthesis to prepare fluorine containing 2,4,5-trisubstituted imidazoles and polysubstituted pyrroles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | >1200°C |
| Melting Point | ∼1170°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Linear Formula | InF3 |
| Merck Index | 14,4959 |
| Solubility Information | Slightly soluble in water. Freely soluble in dilute acids. |
| Formula Weight | 171.82 |
| Percent Purity | 99.95% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.