missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) iodide, 99.998%, (trace metal basis)

Product Code. 10711365 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10711365 25 g Glass bottle 25g
10553421 1 g Glass bottle 1g
10202992 5 g Glass bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10711365

Brand: Thermo Scientific Acros 316010250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13510-35-5 | InI3 | 495.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-35-5
Molecular Formula InI3
Molecular Weight (g/mol) 495.53
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I

Specifications

Melting Point 210.0°C
Color Brown-Yellow to Yellow
Physical Form Crystals or Powder
Quantity 25 g
Assay Percent Range 22.7 to 23.5% (In) (Complexometry)
Linear Formula InI3
Solubility Information Solubility in water: soluble
Formula Weight 495.53
Percent Purity 99.998%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.