missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) iodide, ultra dry, 99.999% (metals basis)

Product Code. 15477385
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477385 2 g 2g
15487385 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477385 Supplier Thermo Scientific Alfa Aesar Supplier No. 047108.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13510-35-5 | I3In | 495.531 g/mol

It is used in the preparation vinyl and methyl indates which undergo regioselective reactions with cinnamyl bromide. It is used as an iodide getter in the Cativa Process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-35-5
Molecular Formula I3In
Molecular Weight (g/mol) 495.531
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I

Specifications

Melting Point 210°C
Color Yellow to Brown
Physical Form Beads
Quantity 2 g
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 4.69 g/mL
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 495.53
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Indium(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.