missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, 99.9997%, (trace metal basis)

Product Code. 11944161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11944161 25 g Glass bottle 25g
11934161 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11944161 Supplier Thermo Scientific Acros Supplier No. 193170250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Color Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 81.0 to 84.3% (In) (Complexometry)
Linear Formula In2O3
Merck Index 15,4992
Solubility Information Solubility in water: insoluble
Formula Weight 277.63
Percent Purity 99.9997%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.