missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfide, 99.995% (metals basis)

Product Code. 11396239
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396239 10 g 10g
11306249 2 g 2g
11386239 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396239 Supplier Thermo Scientific Alfa Aesar Supplier No. 044836.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12030-24-9 | In2S3 | 325.82 g/mol

Indium(III) sulfide emerged as a promising low-hazard buffer material and is used to improve the properties of solar cells and also replaces the hazardous CdS. It is a mid band gap semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-24-9
Molecular Formula In2S3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011061
InChI Key SIXIBASSFIFHDK-UHFFFAOYSA-N
IUPAC Name diindium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[In+3].[In+3]

Specifications

Melting Point 1050°C
Color Red to Orange
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water and organic solvents.
Formula Weight 325.83
Percent Purity 99.995%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Indium (III) sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.