missing translation for 'onlineSavingsMsg'
Learn More

Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)

Product Code. 11355407
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11355407

Brand: Thermo Scientific Alfa Aesar 042677.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 50926-11-9 | InOSn | 249.527 g/mol

Indium tin oxide is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]

Specifications

Physical Form Pieces
Quantity 100 g
Solubility Information Insoluble in water.
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Indium tin oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.