missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Acids and Alkalis

Product Code. 15349025 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
250 mm
355 mm
450 mm
Height (Metric):
26 mm
37 mm
52 mm
Unit Size:
1 roll
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
15349025 450 mm 52 mm 1 roll
15329025 250 mm 26 mm 1 roll
15339025 355 mm 37 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15349025 Supplier Brady™ Supplier No. N007118
  • ONLINE EXCLUSIVE PRICE
    182.00 €
    172.60 € / 1 rolls
    Save 9.40 €
    5% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Acids and Alkalis - Alkali
Adhesive Type Permanent Acrylic, Pressure-sensitive
Coating Silicone
Color White on Purple
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Alkali
For Use With (Application) Pipe and valve marking
Languages English
No. per Pack 30 Markers/Roll
Quantity 30 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 450 mm
Length (English) 36.09 yd.
Length (Metric) 33 m
Width (English) 17.72 in.
Width (Metric) 450 mm
Height (English) 2.05 in.
Height (Metric) 52 mm
Height (Metric) Character 22.5 mm
Alternate Part Number ALKALI 450X52RL-T1-LL-P21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.