missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Air

Product Code. 15359205
Change view
Click to view available options
Display Text:
Air
Breathing air
Compressed air
Hot air
Instrument air
Height (Metric):
26 mm
37 mm
52 mm
Width (Metric):
250 mm
355 mm
450 mm
Unit Size:
1 roll
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Height (Metric) Width (Metric) unitSize
15359205 Air 26 mm 250 mm -
15329235 Breathing air 26 mm 250 mm 1 roll
15379205 Air 52 mm 450 mm -
15339235 Breathing air 37 mm 355 mm 1 roll
15349235 Breathing air 52 mm 450 mm 1 roll
15379215 Compressed air 26 mm 250 mm 1 roll
15389215 Compressed air 37 mm 355 mm 1 roll
15399215 Compressed air 52 mm 450 mm -
15319215 Hot air 26 mm 250 mm -
15329215 Hot air 37 mm 355 mm 1 roll
15339215 Hot air 52 mm 450 mm -
15369225 Instrument air 26 mm 250 mm -
15379225 Instrument air 37 mm 355 mm 1 roll
15389225 Instrument air 52 mm 450 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 15359205 Supplier Brady™ Supplier No. N008298
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Air - Air
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Coating Silicone
Color White on Blue
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Air
For Use With (Application) Pipe and valve marking
Languages English, French
No. per Pack 30 Markers/Roll
Quantity 30 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 250 mm
Length (English) 36.09 yd.
Length (Metric) 33 m
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (English) 1.02 in.
Height (Metric) 26 mm
Height (Metric) Character 13.3 mm
Alternate Part Number AIR 250X26RL-T1-LL-P8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.