missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Fire Fighting

Product Code. 15389245 Shop All Brady Products
Change view
Click to view available options
Display Text:
Fire protection water
Foam
Sprinkler water
Height (Metric):
26 mm
37 mm
52 mm
Width (Metric):
250 mm
355 mm
450 mm
Unit Size:
1 roll
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Height (Metric) Width (Metric) Unit Size
15389245 Fire protection water 26 mm 250 mm 1 roll
15309255 Fire protection water 37 mm 355 mm -
15319255 Fire protection water 52 mm 450 mm 1 roll
15329265 Foam 37 mm 355 mm 1 roll
15319265 Foam 26 mm 250 mm -
15339265 Foam 52 mm 450 mm -
15369255 Sprinkler water 37 mm 355 mm -
15359255 Sprinkler water 26 mm 250 mm -
15379255 Sprinkler water 52 mm 450 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15389245 Supplier Brady™ Supplier No. N008532
  • ONLINE EXCLUSIVE PRICE
    120.00 €
    114.10 € / 1 rolls
    Save 5.90 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Fire Fighting - Fire protection water
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Coating Silicone
Color White on Red
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Fire protection water
For Use With (Application) Pipe and valve marking
Languages English
No. per Pack 30 Markers/Roll
Quantity 30 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 250 mm
Length (English) 36.09 yd.
Length (Metric) 33 m
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (English) 1.02 in.
Height (Metric) 26 mm
Height (Metric) Character 13.3 mm
Alternate Part Number FIRE PROTECTION WATER 250X26RL-T1-LL-P04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.