missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Flammable/Non-Flammable Liquids/Oils
Shop All Brady ProductsDescription
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
Specifications
| Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC |
| Description | Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Flammable/Non-Flammable Liquids/Oils - Lubricating oil |
| Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
| Coating | Silicone |
| Color | White on Dark Brown |
| Format | Roll |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Lubricating oil |
| For Use With (Application) | Pipe and valve marking |
| Show More |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?