missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads, with Pictograms - Gas

Product Code. 15399035 Shop All Brady Products
Change view
Click to view available options
Display Text:
Argon
Bio gas
CO2
Carbon dioxide
Condensate
Exhaust
Freon
Freon 22
Natural gas
Nitrogen
Height (Metric):
26 mm
37 mm
52 mm
Width (Metric):
250 mm
355 mm
450 mm
Unit Size:
1 roll
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Height (Metric) Width (Metric) Unit Size
15399035 Nitrogen 26 mm 250 mm 1 roll
15369045 Bio gas 26 mm 250 mm -
15379045 Bio gas 37 mm 355 mm 1 roll
15389045 Bio gas 52 mm 450 mm 1 roll
15369115 Natural gas 26 mm 250 mm 1 roll
15379115 Natural gas 37 mm 355 mm -
15309035 Argon 26 mm 250 mm -
15319035 Argon 37 mm 355 mm 1 roll
15389115 Natural gas 52 mm 450 mm 1 roll
15339035 Argon 52 mm 450 mm 1 roll
15309105 Carbon dioxide 37 mm 355 mm 1 roll
15319105 Carbon dioxide 52 mm 450 mm 1 roll
15389095 Carbon dioxide 26 mm 250 mm 1 roll
15339055 CO2 26 mm 250 mm -
15349055 CO2 37 mm 355 mm -
15359055 CO2 52 mm 450 mm 1 roll
15319065 Condensate 26 mm 250 mm 1 roll
15339065 Condensate 37 mm 355 mm -
15389065 Exhaust 26 mm 250 mm 1 roll
15349065 Condensate 52 mm 450 mm -
15349085 Freon 22 26 mm 250 mm 1 roll
15309075 Exhaust 37 mm 355 mm -
15319075 Exhaust 52 mm 450 mm 1 roll
15359085 Freon 22 37 mm 355 mm -
15369085 Freon 22 52 mm 450 mm -
15359075 Freon 26 mm 250 mm 1 roll
15379075 Freon 52 mm 450 mm -
15369075 Freon 37 mm 355 mm 1 roll
15329045 Nitrogen 52 mm 450 mm 1 roll
15319045 Nitrogen 37 mm 355 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
This item is not returnable. View return policy
Product Code. 15399035 Supplier Brady™ Supplier No. N007357
  • ONLINE EXCLUSIVE PRICE
    120.00 €
    114.10 € / 1 rolls
    Save 5.90 €
    4% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Individual linerless Pipe Markers on a Roll with die-cut arrowheads, with pictograms - Gas - Nitrogen
Adhesive Type Permanent Acrylic, Pressure-sensitive
Coating Silicone
Color Black on Ochre
Display Warning
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Nitrogen
For Use With (Application) Pipe and valve marking
Languages English
No. per Pack 30 Markers/Roll
Quantity 30 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 250 mm
Length (English) 36.09 yd.
Length (Metric) 33 m
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (English) 1.02 in.
Height (Metric) 26 mm
Height (Metric) Character 13 mm
Alternate Part Number NITROGEN 250X26RL-T1-LL-P15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.