missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Water (A-H)

Product Code. 15399275 Shop All Brady Products
Change view
Click to view available options
Display Text:
Canal water
Chilled water
Chlorated water
Cold water return
Cold water supply
Condensate
Cooling water
Demineralised Water
Drinking water
Heating return
Heating supply
Hot water
Hot water return
Hot water supply
Height (Metric):
26 mm
37 mm
52 mm
Width (Metric):
250 mm
355 mm
450 mm
Unit Size:
1 roll
42 product options available for selection
Product selection table with 42 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Height (Metric) Width (Metric) Unit Size
15399275 Heating supply 26 mm 250 mm -
15359405 Canal water 37 mm 355 mm -
15349405 Canal water 26 mm 250 mm -
15369405 Canal water 52 mm 450 mm 1 roll
15369455 Chilled water 26 mm 250 mm 1 roll
15389455 Chilled water 52 mm 450 mm -
15379455 Chilled water 37 mm 355 mm 1 roll
15399335 Chlorated water 37 mm 355 mm 1 roll
15389335 Chlorated water 26 mm 250 mm 1 roll
15309345 Chlorated water 52 mm 450 mm -
15379415 Cold water return 26 mm 250 mm -
15389415 Cold water return 37 mm 355 mm -
15309415 Cold water supply 26 mm 250 mm -
15399415 Cold water return 52 mm 450 mm -
15319415 Cold water supply 37 mm 355 mm -
15329415 Cold water supply 52 mm 450 mm 1 roll
15359295 Condensate 26 mm 250 mm -
15369295 Condensate 37 mm 355 mm -
15379295 Condensate 52 mm 450 mm 1 roll
15339365 Cooling water 26 mm 250 mm 1 roll
15349365 Cooling water 37 mm 355 mm 1 roll
15359365 Cooling water 52 mm 450 mm -
15329385 Demineralised Water 26 mm 250 mm -
15339385 Demineralised Water 37 mm 355 mm 1 roll
15349385 Demineralised Water 52 mm 450 mm -
15319445 Drinking water 26 mm 250 mm -
15329445 Drinking water 37 mm 355 mm 1 roll
15339445 Drinking water 52 mm 450 mm 1 roll
15379285 Heating return 52 mm 450 mm -
15369285 Heating return 37 mm 355 mm -
15359285 Heating return 26 mm 250 mm -
15309285 Heating supply 37 mm 355 mm 1 roll
15319285 Heating supply 52 mm 450 mm -
15329335 Hot water return 26 mm 250 mm 1 roll
15339335 Hot water return 37 mm 355 mm 1 roll
15349335 Hot water return 52 mm 450 mm -
15369325 Hot water supply 26 mm 250 mm -
15379325 Hot water supply 37 mm 355 mm 1 roll
15389325 Hot water supply 52 mm 450 mm -
15349325 Hot water 37 mm 355 mm 1 roll
15339325 Hot water 26 mm 250 mm 1 roll
15359325 Hot water 52 mm 450 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
42 options
This item is not returnable. View return policy
Product Code. 15399275 Supplier Brady™ Supplier No. N008613
  • ONLINE EXCLUSIVE PRICE
    120.00 €
    114.10 € / 1 rolls
    Save 5.90 €
    4% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Water - Heating supply
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Coating Silicone
Color White on Green
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Heating supply
For Use With (Application) Pipe and valve marking
Languages English
No. per Pack 30 Markers/Roll
Quantity 30 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 250 mm
Length (English) 36.09 yd.
Length (Metric) 33 m
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (English) 1.02 in.
Height (Metric) 26 mm
Height (Metric) Character 13.3 mm
Alternate Part Number HEATING SUPPLY 250X26RL-T1-LL-P03
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.