missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Water (I-Z)

Product Code. 15399445 Shop All Brady Products
Change view
Click to view available options
Display Text:
Industrial water
Non-drinking water
Process water
Raw water
Return
Soft water
Supply
Treated water
Waste water
Water
Well water
Height (Metric):
26 mm
37 mm
52 mm
Width (Metric):
250 mm
355 mm
450 mm
Unit Size:
1 roll
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Height (Metric) Width (Metric) Unit Size
15399445 Process water 52 mm 450 mm -
15339425 Industrial water 26 mm 250 mm -
15349425 Industrial water 37 mm 355 mm 1 roll
15359425 Industrial water 52 mm 450 mm -
15349435 Non-drinking water 52 mm 450 mm -
15329435 Non-drinking water 26 mm 250 mm 1 roll
15379445 Process water 26 mm 250 mm 1 roll
15339435 Non-drinking water 37 mm 355 mm 1 roll
15389445 Process water 37 mm 355 mm -
15379315 Raw water 26 mm 250 mm -
15389315 Raw water 37 mm 355 mm 1 roll
15399315 Raw water 52 mm 450 mm 1 roll
15309485 Return 26 mm 250 mm -
15319485 Return 37 mm 355 mm -
15329485 Return 52 mm 450 mm -
15339395 Soft water 37 mm 355 mm 1 roll
15329395 Soft water 26 mm 250 mm -
15349395 Soft water 52 mm 450 mm -
15309275 Supply 26 mm 250 mm 1 roll
15329275 Supply 52 mm 450 mm -
15319275 Supply 37 mm 355 mm -
15359465 Treated water 26 mm 250 mm 1 roll
15369465 Treated water 37 mm 355 mm 1 roll
15379465 Treated water 52 mm 450 mm -
15319475 Waste water 26 mm 250 mm -
15329475 Waste water 37 mm 355 mm -
15339475 Waste water 52 mm 450 mm -
15349305 Water 26 mm 250 mm 1 roll
15359305 Water 37 mm 355 mm 1 roll
15369305 Water 52 mm 450 mm 1 roll
15379355 Well water 26 mm 250 mm -
15389355 Well water 37 mm 355 mm -
15399355 Well water 52 mm 450 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
This item is not returnable. View return policy
Product Code. 15399445 Supplier Brady™ Supplier No. N009317
  • ONLINE EXCLUSIVE PRICE
    182.00 €
    172.60 € / 1 rolls
    Save 9.40 €
    5% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Water - Process water
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Coating Silicone
Color White on Green
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Process water
For Use With (Application) Pipe and valve marking
Languages English
No. per Pack 30 Markers/Roll
Quantity 30 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 450 mm
Dimensions (L x W x H) 33 m x 450 mm x 52 mm
Length (English) 36.09 yd.
Length (Metric) 33 m
Width (English) 17.72 in.
Width (Metric) 450 mm
Height (English) 2.05 in.
Height (Metric) 52 mm
Height (Metric) Character 22.5 mm
Alternate Part Number PROCESS WATER 450X52RL-T1-LL-P03
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.