missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, with Pictograms -- Flammable/Non Flammable Liquids/Oils

Product Code. 15379195 Shop All Brady Products
Change view
Click to view available options
Display Text:
Alcohol
Diesel
Diesel Fuel
Fuel
Fuel return
Gasoline
White Spirit
Unit Size:
Pack of 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
15379195 White Spirit Pack of 3
15349135 Alcohol -
15339165 Diesel -
15379155 Diesel Fuel -
15379145 Fuel -
15349145 Gasoline Pack of 3
15349155 Fuel return -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15379195 Supplier Brady™ Supplier No. N008274
  • ONLINE EXCLUSIVE PRICE
    17.05 €
    16.31 € / Pack of 3
    Save 0.74 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Flammable/Non Flammable Liquids/Oils - White Spirit
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Dark Brown
Display Danger
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text White Spirit
For Use With (Application) Pipe and valve marking
Labels per Sheet 3
Languages English
No. per Pack 3 Markers/Card
Quantity 3 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 37 x 335 mm
Width (English) 13.98 in.
Width (Metric) 355 mm
Height (English) 1.46 in.
Height (Metric) 37 mm
Height (Metric) Character 16.5 mm
Alternate Part Number WHITE SPIRIT 355X37CARD-T1-P19
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.