missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, with Pictograms, Gas

Product Code. 15399125 Shop All Brady Products
Change view
Click to view available options
Display Text:
Argon
Bio gas
CO2
Carbon dioxide
Condensate
Cooling gas
Exhaust
Freon
Freon 11
Freon 22
Gas
Helium
Inert gas
LPG
Mixed gas
Natural gas
Nitrogen
Recycled gas
Unit Size:
Pack of 3
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
15399125 Helium -
15359045 Bio gas -
15379025 Argon Pack of 3
15329055 CO2 -
15359095 Carbon dioxide Pack of 3
15309065 Condensate -
15359105 Cooling gas -
15369065 Exhaust Pack of 3
15349075 Freon -
15309085 Freon 11 -
15309095 Gas -
15329085 Freon 22 -
15389105 Inert gas -
15369125 LPG Pack of 3
15319115 Mixed gas Pack of 3
15359115 Natural gas -
15389035 Nitrogen Pack of 3
15319125 Recycled gas Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 15399125 Supplier Brady™ Supplier No. N007660
  • ONLINE EXCLUSIVE PRICE
    17.05 €
    16.31 € / Pack of 3
    Save 0.74 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Helium
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color Black on Ochre
Display Warning
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Helium
For Use With (Application) Pipe and valve marking
Labels per Sheet 3
Languages English
No. per Pack 3 Markers/Card
Quantity 3 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 37 x 335 mm
Width (English) 13.98 in.
Width (Metric) 355 mm
Height (English) 1.46 in.
Height (Metric) 37 mm
Height (Metric) Character 16.5 mm
Alternate Part Number HELIUM 355X37CARD-T1-P15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.